Skip to Content
Merck
All Photos(1)

Key Documents

AB5252P

Sigma-Aldrich

Anti-Glial Derived Neurotrophic Factor Antibody

Chemicon®, from sheep

Synonym(s):

GDNF

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$614.00

$614.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$614.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$614.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GDNF(2668)

Application

Detect Glial Derived Neurotrophic Factor using this Anti-Glial Derived Neurotrophic Factor Antibody validated for use in IH.
Immunohistochemistry: 1 μg/mL

Immunoblotting: 1 μg/mL using TBS (not PBS) and blocking is best performed with IgG free BSA highest quality. Antibody recognizes ~75 kDa band in non-acid-treated serum samples corresponding to the GDNF/GDNFR complex. When samples are acid treated, a prominent 32 kDa (dimer) and weaker 16 kDa monomer are observed. Acid treatment: homogenized tissue samples in PBS are acid-treated by adding 1N HCL until the pH is less than 3.0 (pH paper strips), incubated 15 minutes at room temperature and then neutralized by adding 1N NaOH to pH 7.6.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any residue. Contains no preservative. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saurabh Khasnavis et al.
The Journal of biological chemistry, 288(29), 20843-20855 (2013-06-08)
Although Parkinson disease (PD) is a progressive neurodegenerative disorder, available animal models do not exhibit irreversible neurodegeneration, and this is a major obstacle in finding out an effective drug against this disease. Here we delineate a new irreversible model to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service