Skip to Content
Merck
All Photos(1)

Key Documents

AB5252

Sigma-Aldrich

Anti-Glial Derived Neurotrophic Factor Antibody

serum, Chemicon®

Synonym(s):

GDNF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$609.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GDNF(2668)

Specificity

Specific for GDNF. No cross reactivity with Neurturin.

Immunogen

Recombinant human GDNF.

Application

Anti-Glial Derived Neurotrophic Factor Antibody detects level of Glial Derived Neurotrophic Factor & has been published & validated for use in WB.
Immunoblotting: 1:3,000 using TBS (not PBS) and blocking is best performed with IgG free BSA highest quality. Antibody recognizes ~75 kDa band in non-acid-treated serum samples corresponding to the GDNF/GDNFR complex. When samples are acid treated, a prominent 32 kDa (dimer) and weaker 16 kDa monomer are observed. Acid treatment: homogenized tissue samples in PBS are acid-treated by adding 1N HCL until the pH is less than 3.0 (pH paper strips), incubated 15 minutes at room temperature and then neutralized by adding 1N NaOH to pH 7.6.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain

Physical form

Sheep serum. Lyophilized. Reconstitute with 100 μL of sterile distilled water. Centrifuge to remove any residue. Contains no preservative. Glycerol (1:1) can be added for additional stability.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C (dry) for up to twelve months. After reconstitution maintain at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl G Gottschalk et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(10), 2287-2300 (2021-01-31)
Parkinson's disease (PD) is the most common neurodegenerative movement disorder in humans. Despite intense investigations, effective therapies are not yet available to halt the progression of PD. Gemfibrozil, a Food and Drug Administration-approved lipid-lowering drug, is known to decrease the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service