Skip to Content
Merck
All Photos(1)

Key Documents

AB4073

Sigma-Aldrich

Anti-Farnesyl Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$670.00

$670.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$670.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$670.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Protein isoprenylation is a post-translational modification that affects about 0.5% of cellular proteins and is essential for the biological activity of proteins. Two enzymes catalyze theattachment of two prenyl groups to the sulfhydryl group of carboxyl-terminal cysteine groups. Proteins which are prenylated by these enzymes have a distinct motif at the C-terminal of the protein, C-A1-A2-X (C = Cysteine, A 1 & A2 = aliphatic amino acids). The two enzymes involved in this transfer are farnesyltransferase and geranylgeranyltransferase. These transfer a 15 carbon farnesyl or a 20 carbon geranygeranyl, respectively, from aprenyl-pyrophosphate to the protein. Examples of proteins containing this C-A-A-X motif are members of the Ras small G protein family, the nuclear lamins and the gamma subunit oftrimeric G proteins. Prenylation of proteins is necessary for membrane association of proteins as well as protein-protein interactions and the nature of the linked isoprenyl group can influence the protein interactions, such as the interaction between G proteins and receptors.

Specificity

Recognizes farnesyl motif derived from isoprenylated C-A-A-X sequence. Also cross-reacts with geranylgeranyl motif. Detects KLH as well.

Immunogen

Antibody developed using farnesyl cysteine conjugated to KLH.

Application

Anti-Farnesyl Antibody detects level of Farnesyl & has been published & validated for use in ELISA & IF.
ELISA

Immunofluoresecence

Optimal working dilutions must be determined by end user.
Research Category
Signaling
Research Sub Category
MAP Kinases

Physical form

Format: Purified
Purified immunoglobulin. Sterile liquid in phosphate buffered saline and 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zurab Siprashvili et al.
Nature genetics, 48(1), 53-58 (2015-11-26)
Small nucleolar RNAs (snoRNAs) are conserved noncoding RNAs best studied as ribonucleoprotein (RNP) guides in RNA modification. To explore their role in cancer, we compared 5,473 tumor-normal genome pairs to identify snoRNAs with frequent copy number loss. The SNORD50A-SNORD50B snoRNA
Fei Liu et al.
Human molecular genetics, 24(16), 4648-4659 (2015-06-03)
Retinitis pigmentosa (RP) affects about 1.8 million individuals worldwide. X-linked retinitis pigmentosa (XLRP) is one of the most severe forms of RP. Nearly 85% of XLRP cases are caused by mutations in the X-linked retinitis pigmentosa 2 (RP2) and RPGR.
C Banfi et al.
Communications biology, 4(1), 1109-1109 (2021-09-23)
The research into the pathophysiology of atherosclerosis has considerably increased our understanding of the disease complexity, but still many questions remain unanswered, both mechanistically and pharmacologically. Here, we provided evidence that the pro-oxidant enzyme Prenylcysteine Oxidase 1 (PCYOX1), in the
Tsuyoshi Waku et al.
iScience, 24(10), 103180-103180 (2021-10-21)
Lipids, such as cholesterol and fatty acids, influence cell signaling, energy storage, and membrane formation. Cholesterol is biosynthesized through the mevalonate pathway, and aberrant metabolism causes metabolic diseases. The genetic association of a transcription factor NRF3 with obesity has been
Kazuhiro Shirozu et al.
Shock (Augusta, Ga.), 42(6), 570-577 (2014-07-22)
Acute liver failure (ALF) is a fatal syndrome associated with massive hepatocyte death. There is no cure for ALF except liver transplantation. Protein farnesylation is a lipid modification of cysteine residues that is catalyzed by farnesyltransferase (FTase) and has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service