Skip to Content
Merck
All Photos(1)

Key Documents

AB3516P

Sigma-Aldrich

Anti-Sodium-Calcium Exchanger 1 Antibody

Chemicon®, from rabbit

Synonym(s):

NCX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC8A1(6546)

Specificity

Recognizes rat Sodium Calcium Exchanger 1 (NCX1). The immunogen shows no significant sequence homology with other NCX proteins.

Immunogen

A 23 amino acid peptide sequence within the cytoplasmic domain 3 rat NCX1 (Low et al. 1993; Furman et al. 1993; Lee et al. 1994; Nicoll et al. 1996; Kumoro et al. 1992; Blaustein & Lederer 1999).

Application

Anti-Sodium-Calcium Exchanger 1 Antibody is an antibody against Sodium-Calcium Exchanger 1 for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 μg/mL using ECL.





ELISA: 0.5-1 μg/mL using 1 μg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijayababu M Radhakrishnan et al.
The Journal of biological chemistry, 290(14), 8964-8974 (2015-02-05)
NCX1 is a Na(+)/Ca(2+) exchanger, which is believed to provide a key route for basolateral Ca(2+) efflux in the renal epithelia, thus contributing to renal Ca(2+) reabsorption. Altered mineral homeostasis, including intestinal and renal Ca(2+) transport may represent a significant
Xuehui Wang et al.
Frontiers in pharmacology, 10, 616-616 (2019-07-10)
Background: Neuregulin (NRG-1), an essential stress-mediated paracrine growth factor, has a cardioprotective effect in failing heart. However, the underlying mechanism remains unclear. The role of NRG-1β in heart failure (HF) rats was examined. Methods and Results: Volume-overload HF rat model
The Na+/Ca2+ exchanger-1 mediates left ventricular dysfunction in mice with chronic intermittent hypoxia.
Chen, L; Zhang, J; Hu, X; Philipson, KD; Scharf, SM
Journal of Applied Physiology (1985)
Douglas E Brenneman et al.
Journal of molecular neuroscience : MN, 68(4), 603-619 (2019-05-12)
Treatment with cannabidiol (CBD) or KLS-13019 (novel CBD analog), has previously been shown to prevent paclitaxel-induced mechanical allodynia in a mouse model of chemotherapy-induced peripheral neuropathy (CIPN). The mechanism of action for CBD- and KLS-13019-mediated protection now has been explored
Sharlene M Day et al.
Physiological genomics, 33(3), 312-322 (2008-03-13)
Relaxation abnormalities are prevalent in heart failure and contribute to clinical outcomes. Disruption of Ca2+ homeostasis in heart failure delays relaxation by prolonging the intracellular Ca2+ transient. We sought to speed cardiac relaxation in vivo by cardiac-directed transgene expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service