Skip to Content
Merck
All Photos(1)

Key Documents

AB3314P

Sigma-Aldrich

Anti-Monocarboxylate Transporter 4 Antibody

Chemicon®, from rabbit

Synonym(s):

MCT4

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$926.00

$926.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$926.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$926.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A3(9123)

Specificity

Recognizes rat Monocarboxylate Transporter 4 (MCT4/MOT4/ SLC16A3). The immunogen shows no significant sequence homology with other MCT. MCT4 is prominently expressed in skeletal muscle and other cells with high glycolytic rate such as tumors cells and white blood cells suggesting an important role in lactic acid efflux.

Note: According to new nomenclature, the old MCT3 has now been reclassified as MCT4. Consult Halestrap and Price (1999) Biochem. J. 343:281-299.

SPECIES REACTIVITIES:

Rat. The immunogen sequence is 100% conserved in human and mouse and 78% conserved in chicken. Reactivity with other species has not been confirmed.

Immunogen

A 19 amino acid peptide sequence near the C-terminus of rat/human/mouse MCT4.

Application

Detect Monocarboxylate Transporter 4 using this Anti-Monocarboxylate Transporter 4 Antibody validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-5 mg/mL using ECL.



ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3546P

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieyun Cao et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 20(1), 435-444 (2017-12-19)
Monocarboxylate transporters (MCTs) are involved in the transport of monocarboxylates such as ketone bodies, lactate, and pharmaceutical agents. CD147 functions as an ancillary protein for MCT1 and MCT4 for plasma membrane trafficking. Sex differences in MCT1 and MCT4 have been
A Nagai et al.
Placenta, 31(2), 126-133 (2009-12-22)
Lactate plays an important role as an alternative energy substrate, especially in conditions with a decreased utility of glucose. Proton-coupled monocarboxylate transporters (MCTs) are essential for the transport of lactate, ketone bodies, and other monocarboxylates through the plasma membrane and
Claire Thomas et al.
American journal of physiology. Endocrinology and metabolism, 293(4), E916-E922 (2007-07-05)
This study investigated the effects of high-intensity training, with or without induced metabolic alkalosis, on lactate transporter (MCT1 and MCT4) and sodium bicarbonate cotransporter (NBC) content in rat skeletal muscles. Male Wistar rats performed high-intensity training on a treadmill 5
Dale Bryant et al.
Scientific reports, 10(1), 13763-13763 (2020-08-15)
Mutations in the SNX14 gene cause spinocerebellar ataxia, autosomal recessive 20 (SCAR20) in both humans and dogs. Studies implicating the phenotypic consequences of SNX14 mutations to be consequences of subcellular disruption to autophagy and lipid metabolism have been limited to
Zohra Benyahia et al.
Cancers, 13(3) (2021-02-06)
To survive and proliferate in solid tumors, cancer cells adapt and evolve rapidly in microenvironments where oxygen and substrate bioavailability fluctuates over time and space. This creates metabolic heterogeneity. Cancer cells can further cooperate metabolically, for example by swapping glycolytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service