Skip to Content
Merck
All Photos(1)

Key Documents

8.41672

Sigma-Aldrich

4-Piperidinone monohydrate hydrochloride

for synthesis

Synonym(s):

4-Piperidinone monohydrate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$62.70
50 G
$182.00

$62.70


Estimated to ship on31 May 2025



Select a Size

Change View
10 G
$62.70
50 G
$182.00

About This Item

Empirical Formula (Hill Notation):
C5H9NO · xHCl · yH2O
CAS Number:
Molecular Weight:
99.13 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
255-602-8
NACRES:
NA.22

$62.70


Estimated to ship on31 May 2025


Quality Level

pH

4.0-5.5 (100 g/L in H2O)

mp

94-96 °C

storage temp.

2-30°C

SMILES string

[Cl-].N1CCC(=O)CC1.O.[H+]

InChI

1S/C5H9NO.ClH.H2O/c7-5-1-3-6-4-2-5;;/h6H,1-4H2;1H;1H2

InChI key

SXWRTZOXMUOJER-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 91 °C
Melting range (upper value): ≤ 94 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service