Skip to Content
Merck
All Photos(1)

Key Documents

8.07063

Sigma-Aldrich

Propionyl chloride

for synthesis

Synonym(s):

Propionyl chloride, Propionic acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$58.60
500 ML
$81.80

$58.60


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$58.60
500 ML
$81.80

About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-170-0
NACRES:
NA.22

$58.60


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

106 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

270 °C

potency

823 mg/kg LD50, oral (Rat)

expl. lim.

3.6-11.9 % (v/v)

bp

80 °C/1013 hPa

mp

-94 °C

transition temp

flash point 6.5 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CC

InChI

1S/C3H5ClO/c1-2-3(4)5/h2H2,1H3

InChI key

RZWZRACFZGVKFM-UHFFFAOYSA-N

Application

  • Propionyl Chloride in Anticancer Research: Propionyl chloride is used in the synthesis of novel quinazolinone acetamides evaluated for their anticancer properties (Hakim et al., 2022).
  • Propionyl Chloride in Antibacterial Agents: Used in the synthesis of nonphenolic diarylheptanoids to study their antibacterial activities (Demir et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.057 - 1.059
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service