Skip to Content
Merck
All Photos(1)

Key Documents

8.40088

Sigma-Aldrich

2-Ethylhexyl methacrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

2-Ethylhexyl methacrylate, 2-Methyl-2-propenoic acid-2-ethylhexyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$52.80
250 ML
$159.00

$52.80


Only 0 left in stock


Select a Size

Change View
25 ML
$52.80
250 ML
$159.00

About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-708-6
NACRES:
NA.22

$52.80


Only 0 left in stock

vapor pressure

0.13 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

380 °C

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

0.6 % (v/v)

bp

218 °C/1013 hPa

mp

-50 °C

transition temp

flash point 92 °C

solubility

<0.1 g/L

density

0.88 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O(CC(CCCC)CC)C(=O)C(=C)C

InChI

1S/C12H22O2/c1-5-7-8-11(6-2)9-14-12(13)10(3)4/h11H,3,5-9H2,1-2,4H3

InChI key

WDQMWEYDKDCEHT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.882 - 0.886
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service