Skip to Content
Merck
All Photos(1)

Key Documents

8.00835

Sigma-Aldrich

2-Ethylhexyl acrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

2-Ethylhexyl acrylate, Acrylic acid 2-ethylhexyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$42.60
500 ML
$68.70

$42.60


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$42.60
500 ML
$68.70

About This Item

Linear Formula:
CH2=CHCOOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
184.28
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-080-7
NACRES:
NA.22

$42.60


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

0.12 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

230 °C

potency

4435 mg/kg LD50, oral (Rat)
7522 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-6.0 % (v/v)

bp

229 °C/1013 hPa

mp

-90 °C

transition temp

flash point 86 °C

density

0.887 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC(CCCC)CC)C(=O)C=C

InChI

1S/C11H20O2/c1-4-7-8-10(5-2)9-13-11(12)6-3/h6,10H,3-5,7-9H2,1-2H3

InChI key

GOXQRTZXKQZDDN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.884 - 0.886
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service