Skip to Content
Merck
All Photos(1)

Key Documents

8.22137

Sigma-Aldrich

4-Propyl-2-thiouracil

for synthesis

Synonym(s):

4-Propyl-2-thiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$68.80
100 G
$247.00

$68.80


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
$68.80
100 G
$247.00

About This Item

Empirical Formula (Hill Notation):
C7H10N2OS
CAS Number:
Molecular Weight:
170.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-103-2
NACRES:
NA.22

$68.80


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

solid

potency

1250 mg/kg LD50, oral (Rat)

pH

6-7.5 (20 °C, 5 g/L in H2O, slurry)

mp

216-220 °C

transition temp

flash point 300 °C

solubility

1.1 g/L

bulk density

159 kg/m3

storage temp.

2-30°C

SMILES string

S=[c]1[nH][c](cc([nH]1)CCC)=O

InChI

1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11)

InChI key

KNAHARQHSZJURB-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 216 °C
Melting range (upper value): ≤ 220 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service