Skip to Content
Merck
All Photos(1)

Key Documents

8.21961

Sigma-Aldrich

Benzyltrimethylammonium chloride

for synthesis

Synonym(s):

Benzyltrimethylammonium chloride, Trimethylbenzylammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$37.60
250 G
$82.80

$37.60


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$37.60
250 G
$82.80

About This Item

Linear Formula:
C6H5CH2(CH3)3NCl
CAS Number:
Molecular Weight:
185.69
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-300-3
NACRES:
NA.22

$37.60


Only 0 left in stock

Request a Bulk Order

vapor pressure

<0.0001 hPa ( 20 °C)

Quality Level

form

crystals

potency

500 mg/kg LD50, oral (Rat)

pH

6-8 (20 °C, 100 g/L in H2O)

mp

243 °C

solubility

800 g/L

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Cl-].[N+](Cc1ccccc1)(C)(C)C

InChI

1S/C10H16N.ClH/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H/q+1;/p-1

InChI key

KXHPPCXNWTUNSB-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Muta. 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service