Skip to Content
Merck
All Photos(1)

Key Documents

8.14606

Sigma-Aldrich

Sodium peroxide

for synthesis

Synonym(s):

Sodium peroxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$87.70

$87.70


Estimated to ship on31 May 2025



Select a Size

Change View
100 G
$87.70

About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
MDL number:
UNSPSC Code:
12352304
EC Index Number:
215-209-4
NACRES:
NA.22
grade:
for synthesis
synthesis grade

$87.70


Estimated to ship on31 May 2025


grade

for synthesis
synthesis grade

Quality Level

reaction suitability

reagent type: oxidant

pH

12.8 (20 °C, 100 g/L in H2O)

mp

660 °C

density

2.8 g/cm3 at 20 °C

bulk density

1138 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Analysis Note

Assay (manganometric): ≥ 95.0 %
Identity (ICP-OES): passes test

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service