Skip to Content
Merck
All Photos(2)

Key Documents

8.21078

Sigma-Aldrich

(S)-(-)-α-Terpineol

for synthesis

Synonym(s):

(S)-(-)-α-Terpineol, p-Menth-1-en-8-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$37.50
250 ML
$182.00
1 L
$571.00

$37.50


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$37.50
250 ML
$182.00
1 L
$571.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
233-986-8
NACRES:
NA.22

$37.50


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

4300 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

bp

215-217 °C/1013 hPa

mp

36 °C

transition temp

flash point 90 °C

density

0.935 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC([C@H]1CCC(=CC1)C)(C)C

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3/t9-/m1/s1

InChI key

WUOACPNHFRMFPN-SECBINFHSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Optical rotation α 20/D (c=20 in Ethanol): -102.5 - -97.5 °
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Coelho et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-07)
Knowledge of composition of beverages volatile fraction is essential for understanding their sensory attributes. Analysis of volatile compounds predominantly resorts to gas chromatography coupled with mass spectrometry (GC-MS). Often a previous concentration step is required to quantify compounds found at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service