Skip to Content
Merck
All Photos(3)

Key Documents

133051

Sigma-Aldrich

2-Methyl-1-butanol

≥99%

Synonym(s):

(±)-2-Methyl-1-butanol, Active amyl alcohol, sec-Butylcarbinol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$55.20
1 L
$113.00

$55.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$55.20
1 L
$113.00

About This Item

Linear Formula:
C2H5CH(CH3)CH2OH
CAS Number:
Molecular Weight:
88.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.20


Check Cart for Availability

Request a Bulk Order

vapor density

3 (vs air)

Quality Level

vapor pressure

3 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

725 °F

expl. lim.

10 %

refractive index

n20/D 1.410±0.002 (lit.)

bp

130 °C mmHg (lit.)

solubility

water: slightly soluble 3.6g/a00g at 30 °C
alcohol: miscible
diethyl ether: miscible

density

0.819 g/mL at 20 °C (lit.)
0.815 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCC(C)CO

InChI

1S/C5H12O/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

QPRQEDXDYOZYLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-(-)-2-Methyl-1-butanol is a precursor for the synthesis of chiral liquid crystals[1]. It is a potential new-biofuel[2].

Application

2-Methyl-1-butanol was used in a study on photodeoxygenation of 1,2-benzodiphenylene sulfoxide[3].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

108.5 °F - closed cup

Flash Point(C)

42.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Comprehensive Approach to (S)-(-)-2-Methyl-1-Butanol as a Convenient Precursor for Synthesis of Chiral Liquid Crystals.
Kaszynski P and Jawdosiuk M.
Mol. Cryst. Liq. Cryst., 174(1), 21-37 (1989)
Anthony F Cann et al.
Applied microbiology and biotechnology, 81(1), 89-98 (2008-09-02)
Recent progress has been made in the production of higher alcohols by harnessing the power of natural amino acid biosynthetic pathways. Here, we describe the first strain of Escherichia coli developed to produce the higher alcohol and potential new biofuel
Erik R Abels et al.
Cell reports, 28(12), 3105-3119 (2019-09-19)
Gliomas are primary, diffusely infiltrating brain tumors. Microglia are innate immune cells in the CNS and make up a substantial portion of the tumor mass. Glioma cells shape their microenvironment, communicating with and reprogramming surrounding cells, resulting in enhanced angiogenesis
E Lucien et al.
The Journal of organic chemistry, 66(13), 4576-4579 (2001-06-26)
We report that the photodeoxygenation of 1,2-benzodiphenylene sulfoxide, 1, generates an intermediate capable of oxidizing the solvent benzene to phenol. The reactivity of the intermediate was probed with various substrates (2-methylbutane, chloride ion, and para-substituted aryl sulfides). The intermediate produced
Marianthi Karali et al.
PloS one, 6(7), e22166-e22166 (2011-08-06)
Gene transfer using adeno-associated viral (AAV) vectors has been successfully applied in the retina for the treatment of inherited retinal dystrophies. Recently, microRNAs have been exploited to fine-tune transgene expression improving therapeutic outcomes. Here we evaluated the ability of retinal-expressed

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service