Skip to Content
Merck
All Photos(1)

Key Documents

8.20706

Sigma-Aldrich

3-Indolyl acetate

for synthesis

Synonym(s):

3-Indolyl acetate, Acetic acid 3-indolyl ester, 3-Acetoxyindole, Indoxyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$105.00
5 G
$512.00

$105.00


Estimated to ship on02 May 2025



Select a Size

Change View
1 G
$105.00
5 G
$512.00

About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-154-2
NACRES:
NA.22

$105.00


Estimated to ship on02 May 2025


Quality Level

Assay

≥99.0% (GC)

mp

127-129 °C

SMILES string

[nH]1c2c(c(c1)OC(=O)C)cccc2

InChI

1S/C10H9NO2/c1-7(12)13-10-6-11-9-5-3-2-4-8(9)10/h2-6,11H,1H3

InChI key

JBOPQACSHPPKEP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 127 °C
Melting range (upper value): ≤ 129 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service