Skip to Content
Merck
All Photos(1)

Key Documents

8.20555

Sigma-Aldrich

Hexyl acetate

for synthesis

Synonym(s):

Hexyl acetate, Acetic acid hexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-572-7

vapor pressure

5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

36100 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

171.5 °C/1013 hPa

mp

-80 °C

transition temp

flash point 56 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3

InChI key

AOGQPLXWSUTHQB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.872 - 0.874
Identity (IR): passes test

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service