Skip to Content
Merck
All Photos(1)

Documents

8.20138

Sigma-Aldrich

1,2,4-Benzenetricarboxylic 1,2-anhydride

for synthesis

Synonym(s):

1,2,4-Benzenetricarboxylic 1,2-anhydride, Trimellitic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4O5
CAS Number:
Molecular Weight:
192.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
209-008-0
NACRES:
NA.22

vapor pressure

<1 Pa ( 20 °C)

Quality Level

form

solid

potency

>2730 mg/kg LD50, oral (Rat)
23000 mg/kg LD50, skin (Rabbit)

expl. lim.

1-7 % (v/v)

pH

2 (20 °C, 21 g/L in H2O)

bp

240 °C/19 hPa

mp

164-168 °C

transition temp

flash point 227 °C

density

1.487 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H4O5/c10-7(11)4-1-2-5-6(3-4)9(13)14-8(5)12/h1-3H,(H,10,11)

InChI key

SRPWOOOHEPICQU-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Melting range (lower value): ≥ 164 °C
Melting range (upper value): ≤ 168 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service