Skip to Content
Merck
All Photos(1)

Documents

8.00592

Sigma-Aldrich

Phthalic anhydride

for synthesis

Synonym(s):

Phthalic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O3
CAS Number:
Molecular Weight:
148.12
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-607-5
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

crystals

autoignition temp.

580 °C

potency

1530 mg/kg LD50, oral (Rat)
>3160 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-10.5 % (v/v)

pH

2 (20 °C, 6 g/L in H2O)

bp

285 °C/1013 hPa

mp

131 °C

transition temp

flash point 152 °C

solubility

6 g/L (slow decomposition)

density

1.527 g/cm3 at 20 °C

bulk density

500‑700 kg/m3

storage temp.

no temp limit

InChI

1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H

InChI key

LGRFSURHDFAFJT-UHFFFAOYSA-N

Application

<ul>
<li><strong>Antibacterial and cytotoxic activity of chitosan derivatives:</strong>Phthalic anhydride-modified chitosan derivatives exhibit significant antibacterial and cytotoxic properties, suggesting their potential application in medical and pharmaceutical products (Sousa et al., 2024).</li>
<li><strong>Heterodinuclear Polymerization Catalysts:</strong> Research identified Al(III)/K(I) polymerization catalysts that utilize phthalic anhydride for producing high-quality polyester polyols, offering advancements in polymer synthesis technology (Shellard et al., 2024).</li>
<li><strong>Advanced epoxy vitrimers:</strong> Phthalic anhydride was used in the design and synthesis of advanced epoxy vitrimers with enhanced glass transition temperatures, important for applications requiring high-performance thermoset polymers (Dai et al., 2023).</li>
<li><strong>Sustainable alkyd resins:</strong> The utilization of phthalic anhydride in the production of alkyd resins from bio-based resources represents a significant move towards more sustainable and environmentally friendly coating applications (Dizman and Cerrahoglu Ka and ccedil;akgil, 2023).</li>
</ul>

Analysis Note

Assay (morpholine method): ≥ 98,0 %(m)
Melting range (lower value): ≥ 129 °C
Melting range (upper value): ≤ 133 °C
Identity (IR): conforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

305.6 °F - DIN 51758

Flash Point(C)

152 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service