Skip to Content
Merck
All Photos(1)

Key Documents

8.18330

Sigma-Aldrich

4-Carboxybutyltriphenylphosphonium bromide

for synthesis

Synonym(s):

4-Carboxybutyltriphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$93.30

$93.30


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
$93.30

About This Item

Linear Formula:
(C6H5)3P(CH2)4COOH · Br
CAS Number:
Molecular Weight:
443.31
MDL number:
UNSPSC Code:
12352107
EC Index Number:
241-782-5
NACRES:
NA.22

$93.30


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

203-207 °C

transition temp

flash point 195 °C

bulk density

360‑400 kg/m3

storage temp.

2-30°C

SMILES string

[P+](CCCCC(=O)[O-])(c3ccccc3)(c2ccccc2)c1ccccc1.Br

InChI

1S/C23H23O2P.BrH/c24-23(25)18-10-11-19-26(20-12-4-1-5-13-20,21-14-6-2-7-15-21)22-16-8-3-9-17-22;/h1-9,12-17H,10-11,18-19H2;1H

InChI key

MLOSJPZSZWUDSK-UHFFFAOYSA-N

Analysis Note

Identity (IR): passes test
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 203 °C
Melting range (upper value): ≤ 207 °C

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service