Skip to Content
Merck
All Photos(1)

Key Documents

8.18316

Sigma-Aldrich

(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl- 5-methylpyrazine

for synthesis

Synonym(s):

(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl- 5-methylpyrazine, Schoellkopf-Hartwig-Reagent, Bislactimether

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$223.00

$223.00


Estimated to ship on10 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$223.00

About This Item

Empirical Formula (Hill Notation):
C10H18N2O2
CAS Number:
Molecular Weight:
198.26
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

$223.00


Estimated to ship on10 April 2025


Request a Bulk Order

Quality Level

form

liquid

bp

103-104 °C/21 hPa

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay isomers (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.001 - 1.003
Spec. rotation [α²0/D (undiluted): 60.0 - 65.0 °
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service