Skip to Content
Merck
All Photos(1)

Key Documents

8.16125

Sigma-Aldrich

Benzyl butyrate

for synthesis

Synonym(s):

Benzyl butyrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$60.30
500 ML
$140.00

$60.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$60.30
500 ML
$140.00

About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-105-1
NACRES:
NA.22

$60.30


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

11.97 hPa ( 109 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2330 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

235-242 °C/1013 hPa

transition temp

flash point 107 °C

density

1.01 g/cm3

storage temp.

2-30°C

SMILES string

O(Cc1ccccc1)C(=O)CCC

InChI

1S/C11H14O2/c1-2-6-11(12)13-9-10-7-4-3-5-8-10/h3-5,7-8H,2,6,9H2,1H3

InChI key

VONGZNXBKCOUHB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.005 - 1.010
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service