Skip to Content
Merck
All Photos(1)

Key Documents

8.07494

Sigma-Aldrich

Pyrrolidine

for synthesis

Synonym(s):

Pyrrolidine, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$36.00
250 ML
$76.70
1 L
$265.00

$36.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$36.00
250 ML
$76.70
1 L
$265.00

About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-648-7
NACRES:
NA.22

$36.00


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

65 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

345 °C

potency

433 mg/kg LD50, oral (Rat)

expl. lim.

1.6-10.6 % (v/v)

pH

12.9 (20 °C, 100 g/L in H2O)

bp

86-88 °C/1013 hPa

mp

-63 °C

transition temp

flash point 3 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Cl-].N1CCCC1.[H+]

InChI

1S/C4H9N.ClH/c1-2-4-5-3-1;/h5H,1-4H2;1H

InChI key

FCLZCOCSZQNREK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.859 - 0.861
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service