Skip to Content
Merck
All Photos(1)

Key Documents

8.14998

Sigma-Aldrich

Isobornyl acetate

for synthesis

Synonym(s):

Isobornyl acetate, Acetic acid isobornyl ester, exo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$85.20

$85.20


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$85.20

About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-727-6
NACRES:
NA.22

$85.20


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

0.13 hPa ( 20 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

autoignition temp.

440 °C (DIN 51794)

potency

>10000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

kinematic viscosity

4.53 cSt(40 °C)

bp

215 °C/1013 hPa

transition temp

flash point 89 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O([C@@H]1[C@@]2(C([C@@H](C1)CC2)(C)C)C)C(=O)C

InChI

1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12-/m1/s1

InChI key

KGEKLUUHTZCSIP-JFGNBEQYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.979 - 0.984
Spec. rotation [α²0/D (undiluted): -4.0 - +2.0 °
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service