![Isobornyl acetate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/316/533/ce559f5c-fcaf-471a-9f21-477561f6f3bc/640/ce559f5c-fcaf-471a-9f21-477561f6f3bc.png)
Isobornyl acetate
![Isobornyl acetate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/316/533/ce559f5c-fcaf-471a-9f21-477561f6f3bc/640/ce559f5c-fcaf-471a-9f21-477561f6f3bc.png)
Synonym(s):
Isobornyl acetate
Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing