Skip to Content
Merck
All Photos(2)

Key Documents

8.14472

Sigma-Aldrich

(1S)-(-)-Borneol

for synthesis

Synonym(s):

(1S)-(-)-Borneol, L-Borneol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$97.00
100 G
$139.00
250 G
$250.00

$97.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$97.00
100 G
$139.00
250 G
$250.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
207-353-1
NACRES:
NA.22

$97.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥96.0% (GC)

form

powder

potency

5800 mg/kg LD50, oral (Rat)

mp

205-208 °C

transition temp

flash point 66 °C

storage temp.

2-30°C

SMILES string

O[C@H]1[C@@]2(C([C@@H](C1)CC2)(C)C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8-,10-/m1/s1

InChI key

DTGKSKDOIYIVQL-NQMVMOMDSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Spec. rotation [α²0/D (c=5 in ethanol): -37.0 - -34.0 °
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

150.8 °F

Flash Point(C)

66 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service