Skip to Content
Merck
All Photos(2)

Key Documents

911356

Sigma-Aldrich

OXONE®, monopersulfate compound

free-flowing, Redi-Dri

Synonym(s):

Potassium peroxymonosulfate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$86.50
100 G
$129.00
500 G
$157.00
1 KG
$186.00

$86.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$86.50
100 G
$129.00
500 G
$157.00
1 KG
$186.00

About This Item

Linear Formula:
KHSO5 · 0.5KHSO4 · 0.5K2SO4
CAS Number:
Molecular Weight:
307.38
MDL number:
UNSPSC Code:
12352120
NACRES:
NA.21
grade:
reagent grade
form:
powder

$86.50


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

product line

Redi-Dri

form

powder

quality

free-flowing

concentration

>4.0% (active oxygen, titration by Na2S2O3 )

pH

2-3 (10 g/L in aqueous solution, pH <6.0)

SMILES string

[K+].[K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.O[S+]([O-])([O-])(=O)=O.O[S+]([O-])([O-])(=O)=O

InChI

1S/5K.2H2O5S.2H2O4S/c;;;;;2*1-5-6(2,3)4;2*1-5(2,3)4/h;;;;;2*1H,(H,2,3,4);2*(H2,1,2,3,4)/q5*+1;;;;/p-5

InChI key

HJKYXKSLRZKNSI-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

General description

OXONE®, monopersulfate compound is a potassium triple salt mainly used as a stable, easy to handle and nontoxic oxidant.[1]

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer!

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Dri offers a more convenient and safer way to handle the products.
A rapid, efficient synthesis of oxaziridines from imines using buffered OXONE has been reported.[2] Also used to study fading of an artist′s colorants.[3]
OXONE®, monopersulfate compound may be used as an alternative to transition-metal oxidants for the conversion of aldehydes to carboxylic acids or esters.[4]
Oxidant used for halogenation of α,β-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

OXONE is a registered trademark of E. I. du Pont de Nemours and Company
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–3 of 3 Questions  
  1. Are there any special storage conditions that must be maintained?

    1 answer
    1. No, Redi-Dri™ products can be stored under the same conditions as the equivalent non-Redi-Dri™ products.

      Helpful?

  2. How does Redi-Dri™ help keep salts and buffers free-flowing?

    1 answer
    1. Redi-Dri™ products are specially handled and the bottle is specially designed to ensure that the product will remain dry without adding impurities or additives to the product. Ensuring that you receive the highest quality material that is easy to handle.

      Helpful?

  3. How long will material remain free-flowing after it is opened?

    1 answer
    1. While the exact amount of time your material will remain free-flowing can vary, this largely depends on the ambient storage conditions and how many times it is opened.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service