Skip to Content
Merck
All Photos(1)

Key Documents

8.14389

Sigma-Aldrich

4,5-Difluorophthalic anhydride

for synthesis

Synonym(s):

4,5-Difluorophthalic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$280.00

$280.00


Estimated to ship on30 May 2025



Select a Size

Change View
1 G
$280.00

About This Item

Empirical Formula (Hill Notation):
C8H2F2O3
CAS Number:
Molecular Weight:
184.10
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.22

$280.00


Estimated to ship on30 May 2025


Quality Level

mp

92-96 °C

density

0.975 g/cm3

storage temp.

2-30°C

SMILES string

Fc1cc2c(cc1F)C(=O)OC2=O

InChI

1S/C8H2F2O3/c9-5-1-3-4(2-6(5)10)8(12)13-7(3)11/h1-2H

InChI key

UATBWQQFLABWKR-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 96.0 %
Melting range (lower value): ≥ 94 °C
Melting range (upper value): ≤ 98 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service