Skip to Content
Merck
All Photos(1)

Key Documents

8.10278

Sigma-Aldrich

Dibromomethane

(stabilised) for synthesis

Synonym(s):

Dibromomethane, Methylene bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$58.90
250 ML
$80.00
2.5 L
$612.00

$58.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$58.90
250 ML
$80.00
2.5 L
$612.00

About This Item

Empirical Formula (Hill Notation):
CH2Br2
CAS Number:
Molecular Weight:
173.83
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-824-2
NACRES:
NA.22

$58.90


Check Cart for Availability

Request a Bulk Order

vapor pressure

44 hPa ( 20 °C)

Quality Level

Assay

≥98.5% (GC)

form

liquid

potency

108 mg/kg LD50, oral (Rat)
>4000 mg/kg LD50, skin (Rabbit)

bp

95-97 °C/1013 hPa

mp

-53 °C

solubility

11.7 g/L

density

2.49 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCBr

InChI

1S/CH2Br2/c2-1-3/h1H2

InChI key

FJBFPHVGVWTDIP-UHFFFAOYSA-N

Application

  • Ultraviolet Dosimetry: Dibromomethane facilitated the development of photochromic perovskite nanocrystals, enhancing their application in ultraviolet radiation monitoring and dosimetry, critical for environmental and health safety measures (Chen et al., 2024).
  • Energy Storage Solutions: It was utilized as both solvent and electrophilic cross-linker in the synthesis of highly porous hyper-cross-linked polymers, which are effective in methane storage under high pressures, showcasing its role in advancing material sciences for energy applications (Yang et al., 2024).
  • Halogenated Compound Sensing: Dibromomethane was instrumental in the creation of photoluminescent copper(I) iodide alkylpyridine thin films, used as sensors for volatile halogenated compounds, thus enhancing detection capabilities in chemical safety and environmental monitoring (Jamshidi et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Density (d 20 °C/ 4 °C): 2.485 - 2.500
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Ozone 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service