Skip to Content
Merck
All Photos(1)

Key Documents

8.18153

Sigma-Aldrich

Diiodomethane

(stabilised) for synthesis

Synonym(s):

Diiodomethane, Methylene iodide

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$115.00
50 ML
$192.00
1 L
$3,490.00

$115.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$115.00
50 ML
$192.00
1 L
$3,490.00

About This Item

Empirical Formula (Hill Notation):
CH2I2
CAS Number:
Molecular Weight:
267.84
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-841-5
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
181 °C/1013 hPa (decomposition)

$115.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

Assay

≥98% (GC)

form

liquid

potency

76 mg/kg LD50, oral (Rat)

dilution

(for synthesis)

bp

181 °C/1013 hPa (decomposition)

mp

6 °C

solubility

0.8 g/L

density

3.3 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ICI

InChI

1S/CH2I2/c2-1-3/h1H2

InChI key

NZZFYRREKKOMAT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 3.300 - 3.320
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service