Skip to Content
Merck
All Photos(1)

Documents

810242

Sigma-Aldrich

Bovine Serum Albumin Sulfhydryl Modified

≥98%, USA origin, Tested For IgG, Protein standard: suitable, enzyme immunoassay: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801

product name

Bovine Serum Albumin Sulfhydryl Modified, pack of 100 g

grade

special quality (Specialty Grade)

Quality Level

Assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pack of 100 g

origin

USA origin

Free Sulfhydrl Content content

0.0-1.0 mmol/mol

technique(s)

enzyme immunoassay: suitable
protein quantification: suitable
western blot: suitable

impurities

Tested For IgG

loss

≤5%

pH

6.8-7.5

anion traces

chloride (Cl-): 0.0-6.0 mg/g

cation traces

Na: 0.0-15.0 mg/g

foreign activity

Bluetongue, none detected
Vesicular Stomatitis, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service