Skip to Content
Merck
All Photos(1)

Key Documents

8.06790

Sigma-Aldrich

2-Nitrophenol

for synthesis

Synonym(s):

2-Nitrophenol, o-Nitrophenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$59.20
500 G
$102.00

$59.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$59.20
500 G
$102.00

About This Item

Linear Formula:
2-(NO2)C6H4OH
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-857-5
NACRES:
NA.22

$59.20


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

0.15 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

550 °C

potency

2830 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

bp

215-216 °C/1013 hPa

mp

44 °C

transition temp

flash point 100.5 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1c(cccc1)O

InChI

1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H

InChI key

IQUPABOKLQSFBK-UHFFFAOYSA-N

Application

2-Nitrophenol for synthesis. CAS No. 88-75-5, EC Number 201-857-5.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 44 °C
Melting range (upper value): ≤ 46 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.9 °F - closed cup

Flash Point(C)

100.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service