Skip to Content
Merck
All Photos(1)

Key Documents

8.06193

Supelco

tert-Amyl alcohol

EMPLURA®

Synonym(s):

tert-Amyl alcohol, Amylenehydrate, 2-Methyl-2-butanol, tert-Pentyl alcohol, Dimethyl ethyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$91.60

$91.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
$91.60

About This Item

Linear Formula:
CH3CH2C(CH3)2OH
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-908-9
NACRES:
NA.21
bp:
102 °C/1013 hPa
vapor pressure:
15.5 hPa ( 20 °C)

$91.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

15.5 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

425 °C

potency

5200 mg/kg LD50, oral (Rat)

expl. lim.

1.3-9.6 % (v/v)

dilution

(low-cost alternative, for basic applications)

pH

6.0 (20 °C, 118 g/L in H2O, neutral)

kinematic viscosity

5 cSt(23 °C)

bp

102 °C/1013 hPa

mp

-8.4 °C

transition temp

flash point 20.5 °C

solubility

118 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CC)(C)C

InChI

1S/C5H12O/c1-4-5(2,3)6/h6H,4H2,1-3H3

InChI key

MSXVEPNJUHWQHW-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Development of a green, concise synthesis of nicotinamide derivatives catalysed by Novozym® 435 from Candida antarctica in sustainable continuous-flow microreactors: Highlights the use of tert-Amyl alcohol in enzymatic reactions, offering a greener alternative for synthesizing complex molecules in pharmaceutical research (Sheng et al., 2024).

  • Enzymatic Synthesis of Ascorbyl Palmitate in a Rotating Bed Reactor: Discusses the role of tert-Amyl alcohol in creating stable environments for enzyme-catalyzed reactions, enhancing product yield and process efficiency in biochemical applications (Holtheuer et al., 2023).

  • Effects of magnetic fields on the enzymatic synthesis of naringin palmitate: Investigates how tert-Amyl alcohol can be used to enhance the solubility and reactivity of substrates in biocatalytic processes, critical for industrial biotechnology applications (Zhang et al., 2018).

  • Ethyl esters production catalyzed by immobilized lipases is influenced by n-hexane and ter-amyl alcohol as organic solvents: Explores the effectiveness of tert-Amyl alcohol as a co-solvent in biodiesel production, demonstrating its impact on enzyme activity and stability in biofuel research (Borges et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.808 - 0.809
Identity (IR): passes test

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany
Novozym is a registered trademark of Novozymes A/S

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.9 °F - closed cup

Flash Point(C)

20.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service