Skip to Content
Merck
All Photos(1)

Key Documents

8.03245

Sigma-Aldrich

1,2-Dimethylimidazole

for synthesis

Synonym(s):

1,2-Dimethylimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$74.30

$74.30


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$74.30

About This Item

Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
217-101-2
NACRES:
NA.22

$74.30


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

480 °C

potency

1300 mg/kg LD50, oral (Rat)

pH

12.6 (20 °C, 500 g/L in H2O)

bp

205 °C/1013 hPa

mp

30-40 °C

transition temp

flash point 93 °C

density

0.99 g/cm3 at 40 °C

storage temp.

2-30°C

SMILES string

[n]1(c(ncc1)C)C

InChI

1S/C5H8N2/c1-5-6-3-4-7(5)2/h3-4H,1-2H3

InChI key

GIWQSPITLQVMSG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 30 °C
Melting range (upper value): ≤ 40 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service