Skip to Content
Merck
All Photos(3)

Key Documents

199885

Sigma-Aldrich

4(5)-Methylimidazole

98%

Synonym(s):

1H-4-Methylimidazole, 4(or 5)-Methylimidazole, 4-Methyl-1H-imidazole, 4-Methylimidazole, 5-Methyl-1H-imidazole, 5-Methylimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$103.00
250 G
$233.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$103.00
250 G
$233.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
1453
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

bp

263 °C (lit.)

mp

44-47 °C (lit.)

solubility

H2O: soluble, clear, colorless to yellow (50 mg/mL)

SMILES string

Cc1c[nH]cn1

InChI

1S/C4H6N2/c1-4-2-5-3-6-4/h2-3H,1H3,(H,5,6)

InChI key

XLSZMDLNRCVEIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4(5)-Methylimidazole was quantified in coffee by GC-MS method[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Chan et al.
Archives of toxicology, 82(1), 45-53 (2007-07-11)
4-Methylimidazole (4MI) is used in the manufacture of pharmaceuticals, photographic chemicals, dyes and pigments, cleaning and agricultural chemicals, and rubber. It has been identified as a by-product of fermentation in foods and has been detected in mainstream and side stream
Eric D Watt et al.
Biophysical journal, 101(2), 411-420 (2011-07-20)
Rate-limiting millisecond motions in wild-type (WT) Ribonuclease A (RNase A) are modulated by histidine 48. Here, we incorporate an unnatural amino acid, thia-methylimidazole, at this site (H48C-4MI) to investigate the effects of a single residue on protein motions over multiple
Ileana Elder et al.
Archives of biochemistry and biophysics, 437(1), 106-114 (2005-04-12)
In the site-specific mutant of human carbonic anhydrase in which the proton shuttle His64 is replaced with alanine, H64A HCA II, catalysis can be activated in a saturable manner by the proton donor 4-methylimidazole (4-MI). From 1H NMR relaxivities, we
Shinji Hashimoto et al.
Biochemistry, 45(32), 9660-9667 (2006-08-09)
Ultraviolet resonance Raman (UVRR) spectroscopy has been used to characterize the structure and hydrogen bonding state of the distal histidine (His42) in horseradish peroxidase (HRP) complexed with carbon monoxide (HRP-CO). The HRP-CO - HRP UVRR difference spectrum in D(2)O solution
Satoshi Ueda et al.
Journal of the American Chemical Society, 134(1), 700-706 (2011-12-01)
The completely N(1)-selective Pd-catalyzed arylation of unsymmetric imidazoles with aryl halides and triflates is described. This study showed that imidazoles have a strong inhibitory effect on the in situ formation of the catalytically active Pd(0)-ligand complex. The efficacy of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service