Skip to Content
Merck
All Photos(1)

Key Documents

8.03222

Sigma-Aldrich

Dibutylamine

for synthesis

Synonym(s):

Dibutylamine, N-Butyl-1-butanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$37.50
1 L
$55.10
2.5 L
$105.00
40 KG
$1,090.00

$37.50


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$37.50
1 L
$55.10
2.5 L
$105.00
40 KG
$1,090.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
129.24
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-921-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
160 °C/1013 hPa
vapor pressure:
2.3 hPa ( 20 °C)

$37.50


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

2.3 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

260 °C (DIN 51794)

potency

189 mg/kg LD50, oral (Rat)
770 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6-6.8 % (v/v)

dilution

(for synthesis)

pH

11.1 (20 °C, 1 g/L in H2O)

bp

160 °C/1013 hPa

mp

-62 °C

transition temp

flash point 39 °C (DIN 51755 Part 1)

density

0.76 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCC)CCCC

InChI

1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3

InChI key

JQVDAXLFBXTEQA-UHFFFAOYSA-N

Application

  • Paper Title: APPLICATION OF PHASE-TRANSFER CATALYSIS TO THE REACTION OF ACRYLAMIDE WITH DIBUTYLAMINE IN WATER–HEPTANE SYSTEM Description of application: Explores the kinetics of acrylamide and dibutylamine reactions using phase-transfer catalysis, relevant for chemical engineering studies. (GS Simonian, 2014).
  • Paper Title: Kinetic Study of the Acylation Reaction of Dibutylcarbamyl Chloride and Dibutylamine Description of application: Details the kinetics of dibutylcarbamyl chloride and dibutylamine in producing tetrabutylurea, useful in industrial applications. (YB Zhou et al., 2023).
  • Paper Title: Dibutylamine (DBA): A highly efficient catalyst for the synthesis of pyrano pyrimidine derivatives in aqueous media Description of application: Discusses the catalytic role of dibutylamine in synthesizing pharmacologically relevant compounds. (AR Bhat et al., 2016).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.758 - 0.760
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F - closed cup

Flash Point(C)

40.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service