Skip to Content
Merck
All Photos(1)

Key Documents

1.03092

Supelco

Dithizone

for analysis (1,5-diphenylthiocarbazone) Reag. Ph Eur

Synonym(s):

Dithizone, Diphenylthiocarbazone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$163.00
25 G
$408.00

$163.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$163.00
25 G
$408.00

About This Item

Empirical Formula (Hill Notation):
C13H12N4S
CAS Number:
Molecular Weight:
256.33
MDL number:
UNSPSC Code:
12352127
EC Index Number:
200-454-1
NACRES:
NB.21

$163.00


Estimated to ship on29 May 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

Assay

≥98% (argentometric)

form

solid

mp

168 °C (decomposition)

bulk density

250 kg/m3

λmax

604-607 nm in chloroform

absorbance ratio

605/445 nm ≥2.5 (chloroform, 0.005 g/L)

storage temp.

2-30°C

SMILES string

S=C(NNc2ccccc2)N=Nc1ccccc1

InChI

1S/C13H12N4S/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)

InChI key

UOFGSWVZMUXXIY-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR-spectrum): passes test
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (Chloroform): 604 - 607 nm
Spec. Absorptivity A1%/1cm (λmax.; 0,005 g/l; chloroform): ≥ 1522
Absorption ratio (605 nm / 445 nm;0.005 g/l; choroform): ≥ 2.5
Sulfated ash (600 °C): ≤ 0.2 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service