Skip to Content
Merck
All Photos(2)

Key Documents

8.03090

Sigma-Aldrich

Biphenyl

for synthesis

Synonym(s):

Biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-163-5
NACRES:
NA.22

vapor pressure

0.07 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

autoignition temp.

570 °C

potency

2140 mg/kg LD50, oral (Rat)

expl. lim.

0.7-3.4 % (v/v)

bp

255 °C/1013 hPa

mp

68-70 °C

transition temp

flash point 113 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

C1(C2=CC=CC=C2)=CC=CC=C1

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 70 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service