8.01632
Benzil
for synthesis
Synonym(s):
Benzil, Dibenzoyl
Select a Size
$41.40
Estimated to ship on01 June 2025
Select a Size
About This Item
$41.40
Estimated to ship on01 June 2025
Recommended Products
vapor pressure
1.3 hPa ( 128 °C)
Quality Level
Assay
≥99.0% (GC)
potency
2710 mg/kg LD50, oral (Rat)
bp
346-348 °C/1013 hPa (decomposition)
mp
95 °C
storage temp.
2-30°C
SMILES string
O=C(c2ccccc2)C(=O)c1ccccc1
InChI
1S/C14H10O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10H
InChI key
WURBFLDFSFBTLW-UHFFFAOYSA-N
Analysis Note
Melting range (lower value): ≥ 93 °C
Melting range (upper value): ≤ 97 °C
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
356.0 °F - closed cup
Flash Point(C)
180 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service