Skip to Content
Merck
All Photos(1)

Key Documents

8.02611

Sigma-Aldrich

2-Chloroaniline

for synthesis

Synonym(s):

2-Chloroaniline, o-Chloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(Cl)C6H4NH2
CAS Number:
Molecular Weight:
127.57
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-426-4
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

>500 °C

potency

1000 mg/kg LD50, skin (Rat)

expl. lim.

2.4-14.2 % (v/v)

bp

207-209 °C/1013 hPa

mp

-3 °C

transition temp

flash point 105 °C

solubility

insoluble 5.13 g/L

density

1.21 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1)N

InChI

1S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

AKCRQHGQIJBRMN-UHFFFAOYSA-N

Application

2-Chloroaniline for synthesis. CAS 95-51-2, chemical formula 2-(Cl)C₆H₄NH₂.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.209 - 1.214
Identity (IR): passes test

Signal Word

Danger

Storage Class Code

6.1A - Combustible substances, toxic


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service