Skip to Content
Merck
All Photos(3)

Key Documents

A41801

Sigma-Aldrich

3-(Trifluoromethyl)aniline

≥99%

Synonym(s):

α,α,α-Trifluoro-m-toluidine, 3-Aminobenzotrifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$31.10
100 G
$80.60
500 G
$146.00

$31.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$31.10
100 G
$80.60
500 G
$146.00

About This Item

Linear Formula:
CF3C6H4NH2
CAS Number:
Molecular Weight:
161.12
Beilstein:
387672
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$31.10


Check Cart for Availability

Request a Bulk Order

vapor pressure

0.3 mmHg ( 20 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.480 (lit.)

bp

187 °C (lit.)

mp

5-6 °C (lit.)

density

1.29 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc(c1)C(F)(F)F

InChI

1S/C7H6F3N/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4H,11H2

InChI key

VIUDTWATMPPKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Lenz et al.
Ecotoxicology and environmental safety, 54(2), 157-168 (2003-01-29)
19F nuclear magnetic resonance (NMR) spectroscopy was used as a specific tool to investigate the metabolism of 3-trifluoromethylaniline (3-TFMA) in the earthworm species Eisenia veneta. Exposure was via a filter-paper contact toxicity test using five exposure levels (1000, 100, 10
N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(1), 214-224 (2006-09-02)
In this work, we will report a combined experimental and theoretical study on molecular and vibrational structure of 3-aminobenzotrifluoride. The FT-Raman and Fourier transform infrared spectra of 3-aminobenzotrifluoride (3ABTF) were recorded in the liquid phase. The equilibrium geometry, harmonic vibrational
Hiroaki Imoto et al.
Chemistry, an Asian journal, 10(8), 1698-1702 (2015-06-23)
A mechanochromic luminescent dye based on a simple aminomaleimide skeleton was readily synthesized in a one-pot process. It exhibited an on/off mechanochromic luminescent switching property dependent on external stimuli, unlike a traditional mechanochromic color change. The green emission was turned
The many roles for fluorine in medicinal chemistry.
William K Hagmann
Journal of medicinal chemistry, 51(15), 4359-4369 (2008-06-24)
Anna Bielenica et al.
European journal of medicinal chemistry, 101, 111-125 (2015-06-30)
A total of 31 of thiourea derivatives was prepared reacting 3-(trifluoromethyl)aniline and commercial aliphatic and aromatic isothiocyanates. The yields varied from 35% to 82%. All compounds were evaluated in vitro for antimicrobial activity. Derivatives 3, 5, 6, 9, 15, 24 and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service