Skip to Content
Merck
All Photos(1)

Documents

289973

Sigma-Aldrich

anti-Diphenylglyoxime

97%

Synonym(s):

α-Benzil dioxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(=NOH)C(=NOH)C6H5
CAS Number:
Molecular Weight:
240.26
Beilstein:
2053615
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

237-240 °C (dec.) (lit.)

SMILES string

O\N=C(c1ccccc1)\C(=N\O)c2ccccc2

InChI

1S/C14H12N2O2/c17-15-13(11-7-3-1-4-8-11)14(16-18)12-9-5-2-6-10-12/h1-10,17-18H/b15-13+,16-14+

InChI key

JJZONEUCDUQVGR-WXUKJITCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

anti-Diphenylglyoxime (α-Benzil dioxime) was used as a chelating agent in differential pulse adsorptive stripping voltammetric (DPASV) determination of cobalt in vegetable animal foodstuffs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mehrorang Ghaedi
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(2), 295-301 (2006-07-18)
Highly sensitive and interference-free sensitized spectrophotometric method for the determination of Ni(II) ions is described. The method is based on the reaction between Ni(II) ion and benzyl dioxime in micellar media in the presence of sodium dodecyl sulfate (SDS). The
S Giroussi et al.
The Science of the total environment, 176(1-3), 135-139 (1995-12-22)
A selective and sensitive voltammetric method for the determination of cobalt in vegetable animal foodstuffs is developed. The method is based on the use of alpha-benzil dioxime (alpha-BD) as a chelating agent for differential pulse adsorptive stripping voltammetry (DPASV) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service