Skip to Content
Merck
All Photos(1)

Key Documents

8.00198

Sigma-Aldrich

Hexanoic acid

for synthesis

Synonym(s):

Hexanoic acid, Caproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$40.00
250 ML
$41.00
1 L
$104.00
25 L
$908.00

$40.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$40.00
250 ML
$41.00
1 L
$104.00
25 L
$908.00

About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-550-7
NACRES:
NA.22

$40.00


Estimated to ship on27 May 2025


Request a Bulk Order

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

380 °C

potency

1906 mg/kg LD50, oral (Rat)
586 mg/kg LD50, skin (Rabbit)

pH

4 (20 °C, 1 g/L in H2O)

bp

206 °C/1013 hPa

mp

-3 °C

transition temp

flash point 110 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCC

InChI

1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)

InChI key

FUZZWVXGSFPDMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.926 - 0.927
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service