Skip to Content
Merck
All Photos(1)

Key Documents

72158

Sigma-Aldrich

BacMagic-2 Transfection Kit - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

5 REACTIONS
$2,070.00

$2,070.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 REACTIONS
$2,070.00

About This Item

UNSPSC Code:
41106617
NACRES:
NA.54

$2,070.00


Estimated to ship on26 May 2025


Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

General description

The BacMagic-2 Transfection Kit provides a positive selection for creating baculovirus recombinants in preparation for recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification. Like BacMagic DNA, BacMagic-2 DNA is derived from an AcNPV genome, contains an ORF 1629 deletion which prevents nonrecombinant virus from replicating in insect cells, and contains a deletion of the nonessential chitinase (chiA) gene, to improve production of secreted and membrane-targeted proteins. BacMagic-2 DNA have an additional deletion of the v-cath gene, a cathespsin-like cysteine protease, resulting in significantly improved quality and yield for most target proteins. The BacMagic-2 Transfection Kit contains the BacMagic-2 DNA, Insect GeneJuice® Transfection Reagent, and a positive control plasmid to monitor co-transfection efficiency. In addition, it contains Sf9 cells and BacVector Medium, for optimal transfection performance.
Not available for sale in Japan

Components

  • 0.5 µg : BacMagic-2 DNA
  • 50 µl : Insect GeneJuice Transfection Reagent
  • 2 x 1: mlSf9 Cells
  • 1 L: BacVector Insect Cell Medium
  • 2 µg : Transfection Control Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Storage and Stability

Ship Code: Multiple Storage ConditionsStorage: Multiple storage requirements

Legal Information

GENEJUICE is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service