69825-M
NovaBlue Competent Cells - Novagen
Escherichia coli, rod shaped
Synonym(s):
Competent Cells for Transformation
About This Item
Recommended Products
Product Name
NovaBlue Competent Cells - Novagen, NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids) and recA endA mutations.
biological source
Escherichia coli
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
transformation efficiency: >1×108 cfu/μg
shipped in
dry ice
storage temp.
−70°C
Related Categories
General description
Components
•2 × 0.2 ml5 × 0.2 mlNovaBlue Competent Cells
•2 × 2 ml4 × 2 mlSOC Medium
•10 µl10 µlTest Plasmid
Warning
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Colony PCR reagents and our colony PCR protocol using REDExtract-N-Amp™ PCR ReadyMix™ and JumpStart™ REDTaq® PCR ReadyMix™ reagents.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service