Skip to Content
Merck
All Photos(1)

Key Documents

70043

Sigma-Aldrich

T7Select® 1-2b DNA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$777.00

$777.00


Estimated to ship on20 May 2025



Select a Size

Change View
10 μG
$777.00

About This Item

UNSPSC Code:
41106616
NACRES:
NA.51

$777.00


Estimated to ship on20 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

2-8°C

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7SELECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service