Skip to Content
Merck
All Photos(1)

Key Documents

70018-M

Sigma-Aldrich

T7Select® Biopanning Kit

Matched reagents for selecting binders from T7Select libraries

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,200.00

$1,200.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 KIT
$1,200.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

$1,200.00


Estimated to ship on01 June 2025


Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

General description

Enrichment of T7 phage displaying target peptides or proteins can easily be accomplished with the T7Select® Biopanning Kit. The kit contains reagents to perform biopanning with target ligands using 96-well microtiter plates or similar solid phases. Biopanning procedures can be tested and optimized using the positive and negative controls provided in the kit. The supplied Positive Control Lysate contains a T7Select415-1 recombinant that displays the S•Tag peptide. When combined with the Negative Control Lysate at known ratios, the degree of enrichment of high affinity binding phage can be monitored by a simple plaque lift assay with the provided detection reagents.
Biopanning selection
Biopanning requires both binding and elution conditions that maintain the infectivity of the phage. The stability of the T7 phage particle enables the use of a variety of elution conditions during biopanning. Phage maintain infectivity following treatment with 1% SDS, 5 M NaCl, 4 M urea, 2 M guanidine-HCl, 10 mM EDTA, reducing conditions (up to 100 mM DTT), and a pH range of 4 to 10. The wide range of conditions available for T7Select biopanning expands the range of targets that can be used successfully.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

  • 25 g Blocking Reagent
  • 2 x 50 ml 10X TBST Wash Buffer
  • 10 ml T7 Elution Buffer
  • 0.2 ml BL21 Glycerol Stock
  • 0.2 ml BLT5403 Glycerol Stock
  • 0.2 ml BLT5615 Glycerol Stock
  • 1010 pfu T7Select Positive Control Lysate
  • 1010 pfu T7Select Negative Control Lysate
  • 50 μl S-protein HRP Conjugate
  • 2 ml T7Select S-protein
  • 50 ml SuperSignal Substrate
  • pkg/25 g LOCATOR Luminescent Labels
  • pkg/25 Development Folders

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7SELECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service