Skip to Content
Merck
All Photos(1)

Key Documents

70005-M

Sigma-Aldrich

T7SelectUP Primer

Sign Into View Organizational & Contract Pricing

Select a Size

500 PMOL
$249.00

$249.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 PMOL
$249.00

About This Item

UNSPSC Code:
12352208
NACRES:
NA.54

$249.00


Estimated to ship on28 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

Mr: 6105

Warning

Toxicity: Standard Handling (A)

Sequence

5ʹ - GGAGCTGTCGTATTCCAGTC - 3ʹ

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service