Skip to Content
Merck
All Photos(1)

Key Documents

69522

Sigma-Aldrich

T7•Tag® Monoclonal Antibody

Synonym(s):

T7 Tag Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

monoclonal

species reactivity

mouse

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

dry ice
wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The T7•Tag® Antibody is a mouse monoclonal antibody (IgG2b) directed against the 11 amino acid gene 10 leader peptide expressed by many of the pET translation vectors as well as pSCREEN® and pRSET vectors. Because the peptide is the natural amino terminal end of the T7 major capsid protein, the antibody also recognizes T7 bacteriophage (e.g., the T7Select® Vectors). For increased versatility and sensitivity, this antibody is also available labeled with biotin and as a direct conjugate with alkaline phosphatase or horseradish peroxidase.

Specificity

T7•Tag peptide MetAlaSerMetThrGlyGlyGlnGlnMetGly; N-terminal, C-terminal or internal

Cross-reactivity: Negligible with bacterial, insect or mammalian cell lysates

Sensitivity: 1–10 ng (Western blot developed with chromogenic substrates) < 1 ng (AP or HRP conjugate developed with chemiluminescent substrates, or Biotin conjugate developed with Streptavidin AP and chromogenic substrates). Also suitable for immunoprecipitation and immunohistochemistry (1–3)

Working dilution- 1:5,000–1:10,000

Packaging

Plastic ampoule

Warning

Multiple Toxicity Values, refer to MSDS

Physical form

Stabilized solution of antibody in 50% glycerol; includes Positive Control Extract containing 31.1 kDa target protein

Other Notes

  1. Krek, W., et al. 1994. Cell 78,, 161.
  2. Roberts, S.G.E. and Green, M. (1994) Nature 371, 717–720.
  3. Joneson, T., McDonough, M., Bar-Sag, D., and Van Aelst, L. (1996) Science 274, 1374–1376.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PEPscreen is a registered trademark of Sigma-Aldrich Co. LLC
T7-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
T7SELECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service