Skip to Content
Merck
All Photos(1)

Key Documents

662098

Sigma-Aldrich

UbcH5c Conjugating Enzyme E2, Human, Recombinant, E. coli

Synonym(s):

UBD2D3, Ubiquitin-conjugating enzyme E2 D3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Product 662098 is not currently sold in your country. Contact Technical Service

Assay

≥95% (SDS-PAGE)

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full-length, recombinant, human UbcH5c expressed in E. coli. UbcH5c, an E2 ubiquitin ligase, is highly homologous to the yeast UbC4/5 family of E2 enzymes. UbcH5c, together with UbcH5a and UbcH5b, are the most active class of E2 enzymes in cell extracts and are associated with regulation of transcription factors such as p53, NF-κB3, and c-Fos. The active enzyme is useful in the study of ubiquitin regulation and inhibitor screening.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM HEPES, 50 mM NaCl, 1 mM DTT, 10% glycerol, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Brzovic, P.S., et al. 2006. Cell Cycle5, 2867.
Gehrke, S.G., et al. 2003 Blood.101, 3288.
Nuber, U., And Scheffner, M., 1999. J. Biol. Chem.274, 7576.
Schwarz, S.E., et al. 1998. J. Biol. Chem.273, 12148.
Jensen, J., et al. 1995. J. Biol. Chem.270, 30408.
Scheffner, M., et al. 1994. Proc. Natl. Acad. Sci.91, 8797.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Miyakawa et al.
Nature communications, 13(1), 531-531 (2022-01-29)
Autophagy has been linked to a wide range of functions, including a degradative process that defends host cells against pathogens. Although the involvement of autophagy in HBV infection has become apparent, it remains unknown whether selective autophagy plays a critical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service