23-035-M
UbcH5c Protein, active, 10 µg
Active, full length, recombinant human UbcH5c. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.
Synonym(s):
Ubiquitin-conjugating enzyme E2 D3, Ubiquitin-protein ligase D3, Ubiquitin-conjugating enzyme E2-17 kDa 3, UBE2D3
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
Quality Level
100
300
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable
NCBI accession no.
General description
N-terminal HA, 6His-tagged, recombinant human UbcH5c full length, expressed in E.coli.
Ubiquitin-conjugating (E2) enzymes are characterized by the presence of a highly conserved ubiquitin-conjugating domain which accommodates ATP-activated ubiquitin (Ub) via a covalently linked thioester onto its active-site residue. E2 enzymes act via selective protein-protein interactions with the ubiquitin-activating E1 enzyme and ubiquitin ligase E3 enzymes and are able to differentiate effects on downstream substrates, either with a single Ub molecule or a Ub chain. While E3s are involved in substrate selection, E2s are the main determinants for selection of the lysine to construct Ub chains, which thereby directly control the cellular fate of the substrate. The UbcH5 family (UbcH5a, UbcH5b and UbcH5c) have been shown to be the most active class of E2 enzymes in cell extracts and are associated with the control of a number of proteins including the transcriptional regulators p53, IκBα and HIF-1α.
Application
Ubiquitination Cascade Comoponent: E2
Physical form
Purified using immobilized metal affinity chromatography.
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service